Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead(II) acetate
Lead(II) acetate 5% (w/v) in aqueous solution
Lead(II) acetate 5% (w/v) in aqueous solution
:
Ricca Chemical
Lead(II) acetate 5% (w/v) in aqueous solution
:
Ricca Chemical
:
301-04-2
:
:
(H₃CCOO)₂Pb
:
MFCD00012452
:
325.29 g/mol
:
1616
:
6.1,III
Appearance:
Clear, colorless, odorless
Refractive Index:
Confirmed concentration
8212086.PDF
RC429232_02132007