Order Entry
Puerto Rico
ContactUsLinkComponent
Lead(II) acetate 2% (w/v) in aqueous solution
Lead(II) acetate 2% (w/v) in aqueous solution
Catalog # RC4291-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Lead(II) acetate 2% (w/v) in aqueous solution
Catalog # RC4291-32
Supplier:  Ricca Chemical
Supplier Number:  R4291000-1A
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    32 oz.
  • Cat. No.:
    RC4291-32
  • Supplier no.:
    R4291000-1A

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless

Specification Test Results

About this item

 :
 :325.29 g/mol
UN:1616
ADR:6.1,III
CAS Number:301-04-2
Formula:(H₃CCOO)₂Pb
MDL Number:MFCD00012452