Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
Iodine (iodine-iodide) 0.1563 N
  RC400232
Small Business Enterprise
 :  
undefined
 
 
Iodine (iodine-iodide) 0.1563 N
  RC400232
 :  4002-32
Small Business Enterprise
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    RC400232
  • Supplier no.:
    4002-32

 

 

  • Appearance:
    Clear, dark brown, Iodine odor
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.1563 ± 0.0002 N at 20.0 °C

 

 

 :
 :~ 100 °C
 :~ 0 °C
 :1.0600 g/cm³
 :GHS08
 :GHS07
 :Ambient