Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
Iodine (iodine-iodide) 0.0282 N
 
undefined
 
Iodine (iodine-iodide) 0.0282 N
 

 

  • Appearance:
    Clear, dark brown, Iodine odor
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.02820 ± 0.00002 N at 20.0 °C