Order Entry
Puerto Rico
ContactUsLinkComponent
Hydrochloric acid 0.3743 N
Hydrochloric acid 0.3743 N
Catalog # RC36435
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Hydrochloric acid 0.3743 N
Catalog # RC36435
Supplier:  Ricca Chemical
Supplier Number:  R3643000-20F
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    5 gal.
  • Cat. No.:
    RC36435
  • Supplier no.:
    R3643000-20F

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 723):
    0.3743 ± 0.0001 N at 20.0 °C

Specification Test Results

About this item

 :
ADR:8,II
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324