Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Crystal violet
Crystal violet stock solution 1% (w/v) alcoholic solution
Crystal violet stock solution 1% (w/v) alcoholic solution
:
Ricca Chemical
Crystal violet stock solution 1% (w/v) alcoholic solution
:
Ricca Chemical
:
548-62-9
:
:
C₂₅H₃₀ClN₃
:
MFCD00011750
:
407.99 g/mol
:
3077
:
9,III
Absorbance (traceable to NIST SRM 930):
Conforms to control
Appearance:
Purple, alcohol odor
ghs_R3232000.pdf
VWR GHS Compliance Letter (US)