Order Entry
Puerto Rico
ContactUsLinkComponent
Iron(III) chloride solution R1
Corrosive
Health
Iron(III) chloride solution R1
Catalog # RC3105-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Corrosive
Health
Iron(III) chloride solution R1
Catalog # RC3105-16
Supplier:  Ricca Chemical
Supplier Number:  R3105000-500B
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, amber
  • Size:
    500 ml
  • Cat. No.:
    RC3105-16
  • Supplier no.:
    R3105000-500B

Specifications

Specification Test Results

  • Appearance:
    Clear to slightly hazy, bright orange, odorless
  • Assay at 20 °C (traceable to NIST SRM 136):
    105 ± 2 g/L FeCl3.6H2O

Specification Test Results

About this item

 :
 :162.21 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.1000 g/cm³
UN:2582
ADR:8,III
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005