
Ammonium iron (III) sulfate 8% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7783-83-7
:
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004
:482.2 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.1000 g/cm³
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, reddish brown, odorless
- Assay at 20 °C (traceable to NIST SRM 136):8.0 ± 0.1 % (w/v)