
EDTA disodium salt 0.1% (w/v) in 5% (v/v) alcohol USP Test Solution (TS)
Catalog # RC2664-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC2664-16
- Supplier no.:R2664000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless to slight alcohol odor
- Assay at 20 °C (traceable to NIST SRM 915):1.00 ± 0.01 g/L
Specification Test Results
About this item
:
:372.24 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:0.9905 g/cm³
:GHS08
Storage Temperature:Ambient
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037