Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination

Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination

Health
Irritant
Flammable
Supplier:   Ricca Chemical
Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination
Supplier:  Ricca Chemical
Certificates
Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination
Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
  • CAS Number:95-45-4
  • Formula:CH₃C(=NOH)C(=NOH)CH₃
  • MDL Number:MFCD00002117
  • Molecular Weight:116.12 g/mol
  • Boiling Pt:~ 77 °C
  • Density:0.8000 g/cm³
  • Flash Pt:~ 15.5 °C
  • Storage Temperature:Ambient
  • UN:1987
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear, colorless, alcohol odor
  • Suitability for Use:
    Red precipitate with Nickel
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    120 mL
    Log in for availability
    Log in for availability
    Each (120ml)
    Each (120ml) - $65.07
    $65.07
    Each (120ml)
       $65.07

    Supplier Number: R2570000-120A
  • Poly bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (16oz)
    Each (16oz) - $119.83
    $119.83
    Each (16oz)
       $119.83

    Supplier Number: R2570000-500A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.