Order Entry
Puerto Rico
ContactUsLinkComponent
Crystal violet 0.1% (w/v) in glacial acetic acid indicator for nonaqueous titrations
Crystal violet 0.1% (w/v) in glacial acetic acid indicator for nonaqueous titrations
Catalog # RC2479-4
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Crystal violet 0.1% (w/v) in glacial acetic acid indicator for nonaqueous titrations
Catalog # RC2479-4
Supplier:  Ricca Chemical
Supplier Number:  R2479000-120C
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    4 oz.
  • Cat. No.:
    RC2479-4
  • Supplier no.:
    R2479000-120C

Specifications

Specification Test Results

  • Appearance:
    Violet, strong acetic odor
  • Suitability for Use:
    Purple (Basic) - Blue-Green (Acidic)

Specification Test Results

About this item

CAS Number:548-62-9
 :
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750
 :407.99 g/mol
UN:3077
ADR:9,III