
Cobalt(II) chloride 2% (w/v) in diluted hydrochloric acid USP Test Solution (TS)
Catalog # RC2208-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC2208-16
- Supplier no.:R2208000-500A
Specifications
Specification Test Results
- Appearance:Clear, light red, odorless
- Assay at 20 °C (traceable to NIST SRM 915):2.00 ± 0.02 % (w/v)
Specification Test Results
About this item
:
:237.93 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0100 g/cm³
:GHS08
Storage Temperature:Ambient
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652