ContactUsLinkComponent
Water Hardness Buffer, without Magnesium EDTA, Ricca Chemical Company
Water Hardness Buffer, without Magnesium EDTA, Ricca Chemical Company
Catalog # 16001-288
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Water Hardness Buffer, without Magnesium EDTA, Ricca Chemical Company
Catalog # 16001-288
Supplier:  Ricca Chemical
Supplier Number:  R9201000-1A
Small Business Enterprise
CAS Number:  
Specifications
  • Size:
    1 L
  • Cat. No.:
    16001-288
  • Supplier no.:
    R9201000-1A
Specifications
About this item
Ammonium Hydroxide/Chloride buffer without Magnesium or EDTA. For use in water hardness testing.