Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:PRAS BHI (sterile) tubes
- Size:5 mL per tube; 10 tubes per box; 5 boxes per pack.
- Storage Conditions:Stored at 2 - 25 °C in the dark until needed
- Cat. No.:101975-852
- Supplier no.:BHI-HK
OxyPRAS™ BHI Broth is a nutritious broth made from infusions of brain, heart tissue, and peptones that supply protein and other nutrients necessary to support the growth of microorganisms
OxyPRAS™ BHI Broth is supplied ready for use, and no further preparation is necessary. However, for anaerobe samples the addition of Oxyrase® for Broth or AnaSelect® for Broth is needed to establish and maintain an anaerobic environment.
The Broth is supplemented with Vitamin K1 and Hemin which provide growth factors required by certain obligate anaerobes, and supports growth of fastidious anaerobes. OxyPRAS™ BHI Broth is reduced with cysteine and anaerobically sterilized using the Oxyrase® Enzyme System. Anaerobic sterilization of a reduced medium prevents the formation of undesirable oxidation procducts that interfere with the growth of anaerobes. OxyPRAS™ BHI Broth is cell-free and will not interfere with gram stain interpretation of cultures grown in it. Each 5mL tube is sealed with a screw cap lid to ensure its sterility and ease of use.