ContactUsLinkComponent
TexQ™ Disinfectant, Quaternary, ITW Texwipe
TexQ™ Disinfectant, Quaternary, ITW Texwipe
Catalog # TWTX650
Supplier:  Texwipe
undefined
TexQ™ Disinfectant, Quaternary, ITW Texwipe
Catalog # TWTX650
Supplier:  Texwipe
Supplier Number:  TX650
Specifications
  • Cat. No.:
    TWTX650
  • Supplier no.:
    TX650
Specifications
About this item
TexQ™ Disinfectant is an EPA-registered quaternary ammonium compound (quat) which serves as a one-step cleaner/disinfectant (effective against a broad spectrum of bacteria, virus and fungi, and inhibits the growth of mold and mildew and their odors) when used as directed
TexQ™ Disinfectant is available in two solutions: Ready-to-Use trigger spray(TX650) and Concentrate gallon bottle (TX651).
Meets FDA guideline for medical device bacterial endotoxin limit of not more than 20 EU/device.
Both formulations have 25% more kill claims.
H5N1 Kill claims: 10 minutes
HIV-1 kill claims: 2 minutes
MRSA Kill claims: 10 minutes
EPA Reg. No. 10324-85
Ordering information: Each bottle includes a ready-to-use trigger sprayer and a set of 5 color indicator dots for easy identification.