Order Entry
Puerto Rico
ContactUsLinkComponent
tert-Butyl methyl ether, anhydrous ≥99.8%, DriSolv®, Supelco®
Flammable
Irritant
tert-Butyl methyl ether, anhydrous ≥99.8%, DriSolv®, Supelco®
Catalog # EM-MX0824-7
Supplier:  MilliporeSigma
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether, anhydrous ≥99.8%, DriSolv®, Supelco®
Catalog # EM-MX0824-7
Supplier:  MilliporeSigma
Supplier Number:  MX0824-7
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. No.:
    EM-MX0824-7
  • Supplier no.:
    MX0824-7

Specifications

Specification Test Results

  • Assay (GC):
    99.8% min
  • Appearance:
    Clear liquid, free from particulates
  • Color (APHA):
    10 max
  • Filtered through 0.2 µm filter:
    To pass test
  • Residue after evaporation:
    5ppm max
  • Water (H2O):
    50ppm max

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
EINECS:216-653-1
UN:2398
ADR:3,II
 :Packed in septum-sealed bottle.
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812