ContactUsLinkComponent
Front Access Storage Bin Dispenser, S-Curve Technologies
Front Access Storage Bin Dispenser, S-Curve Technologies
Catalog # 89208-708
undefined
Front Access Storage Bin Dispenser, S-Curve Technologies
Catalog # 89208-708
Supplier Number:  SBD-19 PETG
Specifications
  • Color:
    Clear
  • Description:
    Dispenser with one compartment and front access
  • Cat. No.:
    89208-708
  • W×D×H:
    42×30.5×43 cm (16.5×12×17")
  • Supplier no.:
    SBD-19 PETG
Specifications
About this item
One compartment dispenser made from PETG.
  • PETG is safe with IPA
  • Flame treated edges for a smooth finish
  • Keyholes on back for wall mounting
Designed for large volumes of gloves, shoe covers, bouffants, etc.