Order Entry
Puerto Rico
ContactUsLinkComponent
KIMTECH PURE® W4 Presaturated Wipers, Kimberly-Clark Professional®
KIMTECH PURE® W4 Presaturated Wipers, Kimberly-Clark Professional®
Catalog # 83009-328
Supplier:  Kimberly-Clark
undefined
KIMTECH PURE® W4 Presaturated Wipers, Kimberly-Clark Professional®
Catalog # 83009-328
Supplier:  Kimberly-Clark
Supplier Number:  06070

Specifications

  • Color:
    White
  • Cat. No.:
    83009-328
  • W×L:
    22.9 x 27.9 cm (9 x 11")
  • Supplier no.:
    06070

Specifications

About this item

Low-linting wipers are designed for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.

Wipers are constructed of a unique, 100% polypropylene, nonwoven fabric that is low in particulate matter and low in extractable levels. Soft, durable wipers contain no binders or reinforcing fibers. Ideally suited for one-step application of alcohol to critical surfaces. Pre-saturated with semiconductor grade alcohol - 70% isopropyl alcohol/30% deionized water.

Ordering information: Packaged in an antistatic, resealable pouch. Packaged for cleanrooms with outer layer case liner.