Acetonitrile ≥99.9%, BAKER ANALYZED ULTRA LC/MS™ for LC-MS, tested for UHPLC, J.T.Baker®
JT9853-2
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Pack type:Flint bottle, narrow mouth
- Size:1 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default
- Cat. No.:JT9853-2
- Supplier no.:9853-02
- Assay (CH₃CN):≥99.9 %
- Appearance:Passes Test
- Gradient Elution Test (a.u.) - 254 nm:≤0.0005
- Gradient Elution Test (a.u.) - 210 nm:≤0.002
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Color (APHA):≤10
- Residue after Evaporation:≤0.8 ppm
- Water (H₂O):≤100 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤25.0 ppb
- LC/MS Suitability - Largest Response on ESI- Negative Mode (as 4-Nitrophenol):≤25.0 ppb
- Trace Impurities - Aluminum (Al):≤25.0 ppb
- Trace Impurities - Calcium (Ca):≤25.0 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤5.0 ppb
- Trace Impurities - Copper (Cu):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤5.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤5.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤50.0 ppb
- Trace Impurities - Tin (Sn):≤5.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
:
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:81.6 °C (1013 hPa)
:–45.7 °C
:-45.7 °C
:0.782 g/cm³ (20 °C)
:2 °C (closed cup)
:Ambient
:200-835-2
:1648
:3,II
:Management of Change (MOC) category = N
:01-2119471307-38
:GHS02
:GHS07
:75-05-8
:H₃CCN
:MFCD00001878