Potassium hydroxide, pellets NF, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIALS US
Potassium hydroxide, pellets NF, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Regulated
About this item
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- EINECS:215-181-3
- ADR:8,II
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- UN:1813
- REACH:01-2119487136-33
2 Options Available Below
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- EINECS:215-181-3
- ADR:8,II
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- UN:1813
- REACH:01-2119487136-33
Documents
Certificates
Search by Lot Number3152 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR3152
SDS
spec_JT3152-5
Product specification
Specifications for Product Family
Specification Test Results
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Insoluble Substances:Passes Test
- NF - Content of Potassium:≥59.9 %
- NF - Limit of Sodium:≤1.0 %
- NF - Assay (Total Alkali):≥85.0 %
- NF - Carbonate (as K₂CO₃):≤3.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (as K₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Phosphate (PO₄):≤100 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Sodium (Na):≤1.0 %
- EP/BP - Assay:85.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.050 %
- JP - Heavy Metals (as Pb):≤30 ppm
- JP - Sodium (Na):Passes Test
- JP - Potassium Carbonate (K₂CO₃):≤2.0 %
- JP - Assay (KOH):≥85.0 %
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Poly bottle, blue T/E cap500 gEach(1/Itm/Bx)$219.59Case of 4 (1/ Itm/Bx)$595.07Supplier Number: 3152-01Specifications:MOC: RegulatedMOC: Regulated
- Poly bottle, blue T/E cap2.5 kgEach(1/Itm/Bx)$555.75Case of 4 (1/ Itm/Bx)$1,483.23Supplier Number: 3152-05Specifications:MOC: RegulatedMOC: Regulated
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



