



Mercury(II) sulfate ≥98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:7783-35-9
:
Formula:HgSO₄
MDL Number:MFCD00011047
:296.65 g/mol
Merck Index:13,05914
Melting Pt:850 °C
Density:6.47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-992-5
ADR:6.1,II
:Management of Change (MOC) category = N
UN:1645
:GHS06
:GHS08
:GHS09
:GHS07
Specification Test Results
- Assay (HgSO₄):≥98.0 %
- Residue after Reduction:≤0.020 %
- Chloride (Cl):≤0.003 %
- Nitrate (NO₃):Passes Test
- Trace Impurities - Iron (Fe):≤0.001 %
- Mercurous Mercury (as Hg):≤0.15 %
- Suitability for Chemical Oxygen Demand (COD) Determination:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Number2642 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals