

Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Regulated
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1.18 g/cm³
EINECS:215-185-5
UN:1824
ADR:8,II
:Management of Change (MOC) category = R
:GHS05
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance (Clear, colorless liquid):Passes Test
- Normality:4.9 - 5.1
- Chloride (Cl):≤10 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Iron (Fe):≤2 ppm
- Endotoxin Concentration (EU/mL:≤2
Specification Test Results
Related Information
Certificates
Search by Lot Number0895 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT0895-7