

Dodecamolybdophosphoric acid hydrate, crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Non-Regulated
About this item
CAS Number:51429-74-4
:
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913
:1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1.62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
ADR:8,III
:Management of Change (MOC) category = N
UN:3260
:GHS05
:GHS07
Specification Test Results
- Insoluble Matter:≤0.01 %
- Chloride (Cl):≤0.02 %
- Sulfate (SO₄):≤0.025 %
- Ammonium (NH₄):≤0.01 %
- Calcium (Ca):≤0.02 %
- Heavy Metals (as Pb):≤0.005 %
- Iron (Fe):≤0.005 %
Specification Test Results
Related Information
Certificates
Search by Lot Number0246 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Frequently Bought Together