Corning® Trypsin
Catalog # 45000-658
Supplier: Corning
CAS Number:
Specifications
- Size:100 ml
- Storage Conditions:Frozen
- Shipping Conditions:Frozen
- Enzyme Name:Trypsin
- Formulation:Trypsin 1X, 0.25% Trypsin in HBSS without Calcium and Magnesium
- Cat. No.:45000-658
- Supplier no.:25-050-CI
Specifications
About this item
0.25% Trypsin in HBSS (Hank’s Balanced Salt Solution) without calcium and magnesium.
- Porcine parvovirus tested
- 6×100 ml
Trypsin is an enzymatic reagent that is used to release adherent cells from tissue plates for passaging. This offering contains trypsin at a concentration of 0.25%.