

Ethanol ≥96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP suitable for use as excipient, SAFC®
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:78 °C (1013 hPa)
:–114 °C
:0.805…0.812 g/cm3 (20 °C)
:17 °C
:Ambient
:200-578-6
:1170
:3,II
:GHS02
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Assay (d20°C/4°C) (m/m):92.6 - 95.2 %
- Assay (d20°C/4°C) (V/V):95.1 - 96.9 %
- Assay (d 15°C/15°C) (V/V):95.1 - 96.9 %
- Assay (d15.56°C/15.56°C) (m/m):92.3 - 93.8 %
- Assay (d15.56°C/15.56°C) (V/V):94.9 - 96.0 %
- Identity (IR):conforms
- Appearance:conforms
- Clarity of solution:conforms
- Color of solution:conforms
- Acidity or alkalinity:≤ 30 ppm
- Density (d 20 °C/20 °C):0.805 - 0.812
- Density (d 15 °C/15 °C):0.809 - 0.816
- Density (d 15,56 °C/15,56 °C):0.812 - 0.816
- Specific gravity (d 15 °C/15 °C):0.80872 - 0.81601
- Absorbance (at 240 nm):≤ 0.40
- Absorbance (between 250nm and 260nm):≤ 0.30
- Absorbance (between 270nm and 340nm):≤ 0.10
- The spectrum shows a steadily descending curve with no observable peaks or shoulders:conforms
- Volatile impurities (Acetaldehyde and Acetal):≤ 10 ppm
- Volatile impurities (Benzene):≤ 2 ppm
- Volatile impurities (Methanol):≤ 200 ppm
- Volatile impurities (Total of other impurities):≤ 300 ppm
- Volatile impurities (disregard limit):≤ 9 ppm
- Residual solvents (ICH Q3C):excluded by the manufacturing process
- Evaporation residue:≤ 25 mg/l