Corning® Sodium Bicarbonate, Corning
Supplier: Corning
Certificates
About this item
Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7.2 - 7.4 in a culture environment
Bicarbonate is a weak buffer with that makes maintaining a pH range of 7.2 - 7.6 somewhat difficult, but it is non-toxic, and is a nutrient source.
Specifications
- Storage Temperature:Ambient
Specifications
Related Information
Certificates
Search by Lot NumberCell Culture Media, Molecular Buffers and Reagents