Corning® Miller's LB Broth, Corning
45001-048
: Corning
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Miller's LB Broth
- Environmentally Preferable:
- Storage Temperature:Ambient
- Shipping Temperature:Ambient
- Cat. No.:45001-048
- Volume:1 L
- Supplier no.:46-050-CM
Miller's LB broth is based on the original Lysogeny Broth (LB) media formulation developed by Luria and Bertani, which was designed for the rapid propagation of recombinant E. coli strains.
- 20-fold concentration sodium chloride compared to the original formulation
- Widely used for the growth and maintenance of cells
- Work with plasmid DNA and recombinant protein production
: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)