Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids, Corning
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids, Corning
Catalog # 45000-698
Supplier:  Corning
CAS Number:  
undefined
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids, Corning
Catalog # 45000-698
Supplier:  Corning
Supplier Number:  25-030-CI
CAS Number:  

Specifications

  • Description:
    MEM, Corning® 100 ml MEM amino acids, 50X Solution [-] L-glutamine
  • Storage Temperature:
    Frozen
  • Shipping Temperature:
    Frozen
  • Cat. No.:
    45000-698
  • Volume:
    100 ml
  • Supplier no.:
    25-030-CI

Specifications

About this item

The addition of MEM nonessential amino acids to medium can improve growth and the viability of propagating cells in culture while reducing the biosynthetic burden on these cells

This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.