Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Supplier: Corning
Certificates
About this item
McCoy's 5A medium was originally formulated to meet the culture requirements of Novikoff Hepatoma cells, and has been found to support the indefinite proliferation of Walker256 carcinoma cells along with a wide range of human and rat normal and transformed cells. Originally modified by Iwakata and Grace, this version of McCoy's 5A includes increased levels of inositol and glucose
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
Specifications
- Storage Temperature:Refrigerate
Specifications
Related Information
Certificates
Search by Lot NumberCell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Frequently Bought Together