Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
: Corning
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Low endotoxin - <0.25 EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
The original formulation was used to culture embryonic mouse cells and has since been modified to support a variety of normal and transformed cells, such as primary cultures of mouse and chicken cells.
- Storage Temperature:2...8 °C
3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning Cell Culture Media Quick Reference Guide Updated.pdf Corning Classical Media Formulations H2-2017 Corning glutagro Media and Supplements Flyer
Product Literature
More
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Corning Cell Culture Media.pdf
Promotion Material
Frequently Bought Together