Corning® Dialyzed FBS, Corning
Catalog # 97065-302
Supplier: Corning
Specifications
- Description:FBS, Dialyzed
- Cat. No.:97065-302
- Supplier no.:35-071-CV
Specifications
About this item
As a natural product, fetal bovine serum contains a wide variety of biological molecules and compounds which can influence cell development
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Premium FBS is sourced from U.S. herds
- Dialyzed via a 10,000MW cutoff filter and dialyzed against normal saline
This serum is dialyzed against physiological saline to remove small (less than 10,000 MW) potential contaminants while preserving the biological integrity of the final product. An extra filtration step of 0.1 micron further ensures purity.