Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Corning® 10X TAE Buffer, Liquid, Corning

Corning® 10X TAE Buffer, Liquid, Corning

10X TAE Buffer, Liquid, 1 L
Catalog # 45001-074
Supplier:   Corning
Supplier #:  46-010-CM
Corning® 10X TAE Buffer, Liquid, Corning
Catalog # 45001-074
Supplier:  Corning
CAS Number:  
Corning® 10X TAE Buffer, Liquid, Corning
Corning® 10X TAE Buffer, Liquid, Corning

10X TAE Buffer, Liquid, 1 L

CAS Number:  
Price
$244.00
Log in for availability
Pack of 6 (1l)
Product Details & Documents

Tris-Acetate-EDTA (TAE) buffer is used in DNA agarose gel electrophoresis, both in the agarose gel itself and the running buffer

Linear, double-stranded DNA separates faster in a TAE buffer-based system compared to Tris-Borate-EDTAA (TBE) buffer, but the latter has a higher buffering capacity and is thus more resistant to changes in pH. Use of TAE vs. TBE buffer depends on downstream applications. TAE is recommended for preparative electrophoresis in which the nucleic acid bands being separated are to be used in cloning, and other work requiring enzymatic applications.

Specifications

Specifications

  • Description:
    10X TAE Buffer, Liquid
  • Size:
    1 L
  • Cat. No.:
    45001-074
  • Shipping Temperature:
    Ambient
  • Storage Temperature:
    Ambient

Specifications