ContactUsLinkComponent
Ampere’s Rule Apparatus
Ampere’s Rule Apparatus
Catalog # 470135-006
Supplier:  Wards
undefined
Ampere’s Rule Apparatus
Catalog # 470135-006
Supplier:  Wards
Supplier Number:  PH0839
Specifications
  • Description:
    Ampere's Rule Apparatus
  • Size:
    7.5 x 19 x 7.5 cm
  • Cat. No.:
    470135-006
  • Supplier no.:
    PH0839
Specifications
About this item
Ampere's Rule Apparatus helps users visualize the power of electromagnetism.
  • Heavy Copper Wire
  • Iron Fillings
  • Trace Lines of Force by Sending Current through the Wire
It features a heavy copper wire and iron fillings that show the direction of the field around the vertical portion of the wire. A compass up to 50 mm in diameter can be placed under the horizontal portion of the wire to show the same effect.
By sending current through the wire and noting the directions in which the needle points, you can trace the lines of force around the wire and determine the direction of the current.