IRADECON® Bleach Spray, Decon Labs
89234-816
- Size:950 mL (32 oz.)
- Cat. No.:89234-816
- Packaging:Poly bottle
- Supplier no.:4632
Decon's IRADECON® is a pre-diluted, stabilized bleach (sodium hypochlorite) solution ideal for disinfecting surfaces in labs and production areas
The solution is premixed and stable and is a safer, more convenient alternative than self-made bleach solutions that are inherently unstable and often lose their efficacy in 24 to 48 hours. Consists of 0.525% sodium hypochlorite (10% bleach), which is the strength recommended by CDC and OSHA for inactivation HIV, HBV and other pathogens.