ContactUsLinkComponent
Citrad™ Citric Acid Liquid Cleaner
Citrad™ Citric Acid Liquid Cleaner
Catalog # 89234-814
Supplier:  Decon Labs
   Sustainable Packaging
   Waste Reducing
undefined
Citrad™ Citric Acid Liquid Cleaner
Catalog # 89234-814
Supplier:  Decon Labs
Supplier Number:  4401
   Sustainable Packaging
   Waste Reducing
Specifications
  • Size:
    3.8 L (1 gal.)
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Cat. No.:
    89234-814
  • Packaging:
    Poly bottle
  • Supplier no.:
    4401
Specifications
About this item
Citrad®concentrated acidic detergent is ideal for the removal of salts, scale, and metal-oxide. Also used for metal brightening and pharmaceutical-process equipment. Completely phosphate-free, free-rinsing and biodegradable
  • Completely free-rinsing
  • Biodegradable
Citrad™ concentrated acidic detergent contains a safe, organic acid and is phosphate-free, so it will not interfere with sensitive equipment or testing procedures. This concentrated formulation must be diluted with distilled, deionized, or clean potable water and can be used as a soak, scrub, or ultrasonic cleaner. Use to clean labware, stills, cleanrooms, optical parts, pharmaceutical equipment, and manufacturing equipment. Citrad™ is also effective in removing metal oxides, scale, soil, grease, chemicals, solvents, and other compounds from surfaces and equipment.