- Description:Freeze spray, Flash Freeze
- Size:325 mL (11 oz.)
- Cat. No.:89234-810
- Supplier no.:3511
Flash Freeze™ Rapid Freezing Spray is moisture-free propelled COLD gas used to rapidly cool or freeze samples in scientific laboratories.