ProGuard® Professional Handcream, Decon Labs
Supplier: Decon Labs
Certificates
About this item
Proguard Professional Handcream combines emollients to moisturize dry, cracked skin with a barrier film that seals in the moisture, seals out the problems
- Combines emollients to moisturize dry, cracked skin with a barrier film that seals in the moisture, seals out the problems
- Effective in eliminating the irritation caused by latex gloves
- Does not contain perfumes, oils, greases, petroleum, aloe or cocoa butter
- Compatible with and will not compromise the protection of latex gloves
It is particularly effective in eliminating the irritation caused by wearing latex gloves. Because Proguard does not contain perfumes, oils, greases, petroleum, aloe or cocoa butter, it is completely compatible with and will not compromise the protection of latex gloves.
Frequently Bought Together