Order Entry
Puerto Rico
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) DNase-, RNase-, protease-free
Corrosive
Irritant
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) DNase-, RNase-, protease-free
Catalog # IB07100
Supplier:  IBI Scientific
CAS Number:  
Corrosive
Irritant
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) DNase-, RNase-, protease-free
Catalog # IB07100
Supplier:  IBI Scientific
Supplier Number:  IB07100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. No.:
    IB07100
  • Supplier no.:
    07100

Specifications

About this item

 :
Merck Index:13,06793
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Refrigerator
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505