ContactUsLinkComponent
Metal Luer to Threaded End Adapters, Cadence Science®
Metal Luer to Threaded End Adapters, Cadence Science®
Catalog # 75997-228
Supplier:  Avantor
undefined
Metal Luer to Threaded End Adapters, Cadence Science®
Catalog # 75997-228
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Male Luer Lock to 10-32 Standard Thread
  • Material:
    Plated brass
  • Environmentally Preferable:
  • Cat. No.:
    75997-228
Specifications
About this item
Luer adapters are used in hospitals, laboratories, and biomedical instruments.
  • Precision machined
  • Threaded ends
  • Made from either nickel plated brass or 316 stainless steel
Cadence Science® metal adapters are precision machined and are made from either nickel plated brass or 316 stainless steel.
Standard luer tapers (meets ISO 594 specifications) include Female Luer Lock (FLL), Micro-Mate® Female Luer Lock (MM FLL), Male Luer Lock (MLL), and Male Luer (ML).