Temperature Measurement Apparatus for Hydrogenation/Gas Apparatus, Ace Glass Incorporated
Catalog # 89063-384
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Temperature Measurement Apparatus, Complete
- Cat. No.:89063-384
- Supplier no.:8648-57
Specifications
About this item
This borosilicate glass vessel, offset adapter, coupling and modified top bottle clamp are used with shaker type Parr Hydrogenation/Gas Apparatus I (89058-912) to allow temperature measurement while connected to hydrogen source
For use with three larger vessels only, with #25 Ace-Thred neck.
Ordering information: This complete apparatus comes complete with three borosilicate glass reaction vessels (89063-232, 89063-238, 89063-240), offset adapter (89051-324) with two #7 and one #15 Ace-Thred connections, #7 Ace-Thred nylon bushing with o-ring (89051-128), PTFE coupling (89052-672) #15 to #25 Ace-Thred, top bottle clamp (89058-886), and a 12-pack of ferrules (89075-734) for #7 Ace-Thred.