Order Entry
Puerto Rico
ContactUsLinkComponent
 
HyClone™ FetalClone™ III serum, U.S. Origin, HyClone products (Cytiva)
 :  Cytiva
 
undefined
HyClone™ FetalClone™ III serum, U.S. Origin, HyClone products (Cytiva)
 :  Cytiva
 

 

  • Catalog No:
  • 16777-240
  • 82013-596
  • 76324-892
  • 16777-238
  • 76236-312
  • Origin:
  • US
  • US
  • US
  • US
  • US
  • Endotoxin:
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • Hemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage Temperature:
  • Less than –10 °C
  • Less than –10 °C
  • Less than –10 °C
  • Less than –10 °C
  • Less than –10 °C