Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TMB membrane enhancer
TMB membrane enhancer
TMB membrane enhancer
Catalog # 95059-160
Supplier:
KPL
CAS Number:
SDS
Expand image
TMB membrane enhancer
Catalog # 95059-160
Supplier:
KPL
Supplier Number:
5420-0026
CAS Number:
SDS
Specifications
Size:
40 ml
Cat. No.:
95059-160
Supplier no.:
50-77-01
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation