ContactUsLinkComponent
TMB Membrane Peroxidase Substrate System, KPL
TMB Membrane Peroxidase Substrate System, KPL
Catalog # 95059-158
Supplier:  KPL
CAS Number:  
TMB Membrane Peroxidase Substrate System, KPL
Catalog # 95059-158
Supplier:  KPL
Supplier Number:  5420-0025
CAS Number:  
Specifications
  • Description:
    TMB Membrane Peroxidase Substrate System
  • Size:
    440 ml
  • Cat. No.:
    95059-158
  • Supplier no.:
    50-77-00
Specifications
About this item
TMB (3, 3', 5, 5' - Tetramethylbenzidine) is the most sensitive colorimetric substrate for peroxidase detection
TMB Membrane Peroxidase is supplied as 3 solutions for simple mix-and-use preparation. The substrate may be adapted as a soluble substrate for ELISA by omitting TMB Membrane Enhancer.
It produces a deep blue stable precipitate at the reaction site in the presence of peroxidase-labeled conjugates, enabling detection of as little as 50 pg of peroxidase. It is recommended for membrane assays and dot blots.