Adapter, Bottom Drip, Internally Threaded, with Flow Regulator Valve, Ace Glass Incorporated
For use with chromatography columns, these bottom drip adapters have a flow regulator valve and an internal thread.
- Bottom drip
- Flow regulator valve
- Internal thread
- Available in PTFE or nylon and four different Ace-Thred sizes
Adapters feature a 1/4-inch-28 UNF 2B internal thread that will accept tubing connectors.
: Supplied with one FETFE o-ring and one polyethylene 100 micron filter disc packing support (fits into the top of the adapter). Glass filter discs are also available. Contact your Avantor representative for further information. For valve stem with o-ring only, order 89052-558 (fits all four Ace-Thread sizes). For replacement o-ring 12-pack, order 89052-528. For optional filter disc remover, order 89052-532.