HyClone™ SG-200 Solution, HyClone products (Cytiva)
Catalog # 82024-258
Supplier: Cytiva
CAS Number:
Specifications
- Description:SG-200, (stable dipeptide of L-alanyL-glutamine) 200 mM, in 0.85% NaCl solution
- Size:100 ml
- Cat. No.:82024-258
- Storage temperature:Less than 0 °C
- Volume:Ea.
- Supplier no.:SH30590.01
Specifications
About this item
HyClone SG-200 Solution is a stable dipeptide of L-Alanyl L-Glutamine
Storage conditions: Store at 2° to 8°C.