HyClone™ Classical Liquid Media: Iscove's Modified Dulbeccos Medium (IMDM), HyClone products (Cytiva)
: Cytiva
HyClone Classical Liquid Media: Iscove's Modified Dulbeccos Medium (IMDM) is manufactured using ISO 9001:2000-certified processes.
- Contains 25 mM glucose, 1,0 mM sodium pyruvate, 25 mM HEPES and phenol red
- Hydrated using WFI quality water
- 0,1 μm sterile filtered
- Manufactured using ISO 9001:2000 certified processes
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as raw material components must pass strict Quality Control testing to confirm the appropriate level of quality. Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection.
We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.
- Storage Temperature:2...8 °C
Frequently Bought Together