Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

4-Fluoroquinoline hydrochloride ≥95%

1 g
Catalog # 101942-262
Marketsource
Supplier:   Matrix Scientific
 :  069859-1G
4-Fluoroquinoline hydrochloride ≥95%
Catalog # 101942-262
Supplier:  Matrix Scientific
Marketsource
CAS Number:  1245643-64-4
4-Fluoroquinoline hydrochloride ≥95%
4-Fluoroquinoline hydrochloride ≥95%

1 g

CAS Number:  1245643-64-4
Price
$930.77
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hazard: Irritant

  • CAS Number:1245643-64-4
  • Formula:C₉H₇ClFN
  • MDL Number:MFCD16249527
  • Molecular Weight:183.61 g/mol
Specifications

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.