Order Entry
Puerto Rico
ContactUsLinkComponent
(6,8-Dichloroimidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride
(6,8-Dichloroimidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride
Catalog # 101920-364
Supplier:  Matrix Scientific
CAS Number:  
(6,8-Dichloroimidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride
Catalog # 101920-364
Supplier:  Matrix Scientific
Supplier Number:  060626-500MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 mg
  • Cat. No.:
    101920-364
  • Supplier no.:
    060626-500MG

Specifications

About this item

 :
 :281.53 g/mol
CAS Number:1235438-87-5
Formula:C₉H₇Cl₃N₂O₂
MDL Number:MFCD16040117