2-(2-Chlorophenyl)azepane
500 mg
Catalog # 101850-078
Marketsource
Supplier: Matrix Scientific
: 040738-500MG
2-(2-Chlorophenyl)azepane
Catalog # 101850-078
Supplier: Matrix Scientific
Marketsource
CAS Number: 887360-60-3
Jump to:
Product Details & Documents
Hazard: Irritant
- CAS Number:887360-60-3
- Formula:C₁₂H₁₆ClN
- MDL Number:MFCD07357623
- Molecular Weight:209.72 g/mol
Specifications
Specifications
- Size:500 mg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



