Order Entry
Puerto Rico
ContactUsLinkComponent
5-Bromo-2-chloro-m-xylene ≥97%
5-Bromo-2-chloro-m-xylene ≥97%
Catalog # 101782-124
Supplier:  Matrix Scientific
CAS Number:  
undefined
5-Bromo-2-chloro-m-xylene ≥97%
Catalog # 101782-124
Supplier:  Matrix Scientific
Supplier Number:  001626-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 g
  • Cat. No.:
    101782-124
  • Supplier no.:
    001626-25G

Specifications

Specification Test Results

  • Boiling point:
    131°C/1mm
  • Density:
    1.48

Specification Test Results

About this item

 :
 :219.51
Boiling Pt:131 °C (1mm)
Density:1.48 g/cm³
CAS Number:206559-40-2
Formula:C₈H₈BrCl
MDL Number:MFCD03094622